Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Ultipor Membrane Optimized for Scale Up
Product Code. 17104451
Change view
Click to view available options
0.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17104451 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17104451 Supplier Cytiva (formerly Pall Lab) Supplier No. 4906
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Syringe filter with Ultipor™ VF Grade DV20 membrane validated for virus removal.
  • Suitable for small-scale membrane qualification studies such as bacteriophage or prion (TSE agent) clearance, as well as membrane flow rate, capacity and protein transmission studies
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 0.2 μm
Sterility Sterile