Learn More
Aclonifen-d5 (phenoxy-d5), CDN
Specifications
Specifications
| Chemical Name or Material | Aclonifen D5 (phenyl D5) |
| CAS | 2469279-06-7 |
| Molecular Formula | C12 D5 H4 Cl N2 O3 |
| Quantity | 0.01 g |
| Synonym | Aclonifen D5 (phenyl D5), 2-Chloro-6-nitro-3-(phenoxy-2,3,4,5,6-d5)benzenamine, 2-Chloro-6-nitro-3-(phenoxy-d5)aniline, Aclonifen-d5, Aclonifen-d5 (phenoxy-d5) |
| InChI Formula | InChI=1 S/C12H9ClN2O3/c13-11-10(18-8-4-2-1-3-5-8)7-6-9(12(11)14)15(16)17/h1-7 H,14H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Oc2ccc(c(N)c2Cl)[N+](=O)[O-])c([2 H])c1[2 H] |
| IUPAC Name | 2-chloro-6-nitro-3-(2,3,4,5,6-pentadeuteriophenoxy)aniline |
| Molecular Weight (g/mol) | 269.6953 |
| Formula Weight | 269.0616 g/mol |
| Show More |