missing translation for 'onlineSavingsMsg'
Learn More

ACK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 EUR - 410.00 EUR

Specifications

Antigen ACK1
Concentration 0.13 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16832884
View Documents
Proteintech
14304-1-AP-20UL
20 μL
145.00 EUR
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16822884
View Documents
Proteintech
14304-1-AP-150UL
150 μL
410.00 EUR
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tyrosine kinase. that after binding to CDC42. inhibits both its intrinsic and stimulated GTPase activity. This protein is a tyrosine kinase that binds Cdc42Hs in its GTP-bound form and inhibits both the intrinsic and GTPase-activating protein (GAP).
Specifications

Specifications

ACK1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Rat, Mouse, Human
O54967, Q07912, Q5U2X5
10188, 303882, 51789
ACK1 Fusion Protein Ag5419
Primary
-20°C
TNK2
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ACK, ACK 1, ACK1, Activated CDC42 kinase 1, p21cdc42Hs, TNK2
Tnk2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.