missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14304-1-AP-150UL
This item is not returnable.
View return policy
Description
Tyrosine kinase. that after binding to CDC42. inhibits both its intrinsic and stimulated GTPase activity. This protein is a tyrosine kinase that binds Cdc42Hs in its GTP-bound form and inhibits both the intrinsic and GTPase-activating protein (GAP).Specifications
| ACK1 | |
| Polyclonal | |
| Unconjugated | |
| TNK2 | |
| ACK, ACK 1, ACK1, Activated CDC42 kinase 1, p21cdc42Hs, TNK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10188, 303882, 51789 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O54967, Q07912, Q5U2X5 | |
| Tnk2 | |
| ACK1 Fusion Protein Ag5419 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction