Learn More
Acetylacetone, Sigma-Aldrich™
Product Code. 15858140
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15858140 1 L 1 L
15838140 5 mL 5mL
15848140 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15858140 Supplier Sigma Aldrich Supplier No. 8000231000
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-54-6
Grade Research
Molecular Formula CH3COCH2COCH3
Molecular Weight (g/mol) 100.12
Specifications
Melting Point −23°C (Lit.)
Density 0.975 g/mL at 25°C
Boiling Point 140.4°C
MDL Number MFCD00008787
Quantity 1 L
Synonym ACAC, 2,4-Pentanedione, 2,4-Pentanedione