Learn More
Acetylacetonatobis(ethylene)rhodium(I), 99%
Product Code. 10354162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10354162 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10354162 Supplier Thermo Scientific Acros Supplier No. 363381000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
CAS: 12082-47-2 | C9H15O2Rh | 258.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 12082-47-2
CAS Min % 98.5
CAS Max % 100.0
Melting Point 141.0°C
Color Orange
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H15O2Rh
MDL Number MFCD00015354
Quantity 100 mg
Synonym acetylacetonatobis ethylene rhodium i, bis ethylene 2,4-pentanedionato rhodium i, acetylacetonatobis ethylene rhodium, ethylene; z-4-oxopent-2-en-2-olate; rhodium
InChI Key FLRBEQQDEGBCJS-FGSKAQBVSA-M
SMILES [Rh+].C=C.C=C.C\C([O-])=C\C(C)=O
IUPAC Name ethene;(Z)-4-oxopent-2-en-2-olate;rhodium
Molecular Weight (g/mol) 258.12
PubChem CID 11630270
Formula Weight 258.13
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Acetylacetonatobis(ethylene)rhodium(I)
Show More Show Less
compliance-icons
Product Identifier
  • Acetylacetonatobis(ethylene)rhodium(I)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.