missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™

Product Code. 11497530 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497530 1 mg 1mg
11407540 5 mg 5mg
11417540 10 mg 10mg
11427540 25 mg 25mg
11437540 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11497530 Supplier MP Biomedicals Supplier No. 0210049001
  • ONLINE EXCLUSIVE PRICE
    55.15 EUR
    52.50 EUR / 1mg
    Save 2.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 75520-41-1
Molecular Formula C23H38Li3N7O17P3S
Molecular Weight (g/mol) 830.39
InChI Key MOGXDJCKYPGZRO-JHJDYNLLSA-N
Synonym acetyl coenzyme a trilithium salt
PubChem CID 129893900
IUPAC Name S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium
SMILES [Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O

Specifications

Color White
Assay Percent Range >92%
Quantity 1 mg
Formula Weight 881.1
Physical Form Powder
Chemical Name or Material Acetyl Coenzyme A Trilithium Salt Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.