Learn More
Acetonitrile, CHROMASOLV™, ≥99.9% (GC), for LC-NMR, Solstice
Product Code. 15644720
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
500mL
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15644720 6 x 500 mL Pack of 6
15624720 500 mL 500mL
15614720 1 L 1 L
15634720 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15644720 Supplier Honeywell-Chromasolv Supplier No. 349556X500ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CHROMASOLV™ NMR, for LC-NMR,≥99.9% (GC)
  • Solvents for LC-NMR applications
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
Spezifikation
Color Colorless
Density 0.786g/mL at 25°C
Quantity 6 x 500 mL
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Linear Formula CH3CN
Vapor Pressure 72.8 mmHg (20°C)
UN Number UN1648
Formula Weight 41.05
Percent Purity ≥99.9% (GC)
Chemical Name or Material Acetonitrile
Mehr anzeigen Weniger anzeigen