missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ≥99.5%, Spectrophotometric Grade, Honeywell Riedel-de Haën™

Product Code. 15691450
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15691450 2 L Glass Bottle 2 L
15681450 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15691450 Supplier Honeywell Riedel-de Haën Supplier No. 1546012L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Spectrophotometric Grade,≥99.5%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -41°C
Density 0.786g/mL at 25°C
Quantity 2 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Packaging Glass Bottle
Refractive Index 1.344
Linear Formula CH3CN
Vapor Pressure 72.8mmHg (20°C)
UN Number UN1648
Beilstein 741857
Boiling Point 81.6°C
Formula Weight 41.05g/mol
Percent Purity ≥99.5%
Grade Spectrophotometry
Chemical Name or Material Acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.