missing translation for 'onlineSavingsMsg'
Learn More

Scharlab ACETONITRIL MULTI 2,5L

Brand:  Scharlab AC03332500


ACETONITRIL MULTI 2,5L

Product Code. 11369793

  • ONLINE EXCLUSIVE PRICE
    320.25 EUR
    204.48 EUR / 2.5 L
    Save 115.77 EUR
    36% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetonitrile, Multisolvent HPCL grade ACS UV-VIS, Reag. Ph Eur

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302+H312+H332-Harmful if swallowed, in contact with skin or if inhaled
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.