Learn More
Acetone 2,4-Dinitrophenylhydrazone-d3, TRC
Product Code. 30327314
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327314 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327314 Supplier Toronto Research Chemicals Supplier No. TRCA16385250MG
 Restricted Item
Estimated Shipment: 08-10-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acetone 2,4-Dinitrophenylhydrazone-d3
CAS 259824-57-2
Molecular Formula C9 D3 H7 N4 O4
Quantity 50 mg
Synonym 2-Propanone, (4,6-dinitrophenyl-2,3,5-d3)hydrazone (9CI), Acetone-2,4-dinitrophenyl-3,5,6-d3-hydrazone, Acetone 2,4-Dinitrophenylhydrazone-d3, Acetone-2,4-dinitrophenylhydrazone-D3 (phenyl-D3), Acetone 2,4-Dinitrophenylhydrazone-3,5,6-d3
InChI Formula InChI=1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3/i3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c(c1NN=C(C)C)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2,3,5-trideuterio-4,6-dinitro-N-(propan-2-ylideneamino)aniline
Molecular Weight (g/mol) 241.219
Formula Weight 241.089