missing translation for 'onlineSavingsMsg'
Learn More

Acetone 2,4-Dinitrophenylhydrazone-13C3, TRC

Product Code. 30322874
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322874 10 mg 10mg
30327673 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30322874 Supplier Toronto Research Chemicals Supplier No. TRCA16385310MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetone 2,4-Dinitrophenylhydrazone-13C3
CAS 395075-27-1
Molecular Formula C613C3H10N4O4
Quantity 10 mg
Synonym 2-Propanone-13C3 2-(2,4-Dinitrophenyl)hydrazone, 2-Propanone-13C3 (2,4-Dinitrophenyl)hydrazone, Acetone-13C3 (2,4-Dinitrophenyl)hydrazone, 1-(2,4-Dinitrophenyl)-2-(propan-2-ylidene)hydrazine-13C3, NSC 131450-13C3, NSC 6120-13C3
InChI Formula InChI=1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3/i1+1,2+1,6+1
SMILES [13CH3]/[13C]([13CH3])=N/NC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1
IUPAC Name 2,4-dinitro-N-((1,2,3-13C3)propan-2-ylideneamino)aniline
Molecular Weight (g/mol) 241.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.