Learn More
Acetoin, TRC
Product Code. 30134904
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134904 25 g 25g
30011414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134904 Supplier Toronto Research Chemicals Supplier No. TRCA16377525G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 513-86-0
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
Synonym 2-Butanone, 3-hydroxy-, 3-Hydroxy-2-butanone, (±)-3-Hydroxybutan-2-one, (±)-Acetoin, 1-Hydroxyethyl methyl ketone, 2,3-Butanolone, 2-Hydroxy-3-butanone, 3-Hydroxybutanone, 3-Hydroxyl-2-butanone, 3-Oxo-2-butanol, Acetoin, Acetoine, Acetyl methyl carbinol, DL-Acetoin, Dimethylketol, Methanol, acetylmethyl-, NSC 7609, γ-Hydroxy-β-oxobutane
IUPAC Name 3-hydroxybutan-2-one
SMILES CC(O)C(=O)C
Tekniske data
Quantity 25 g
InChI Formula InChI=1S/C4H8O2/c1-3(5)4(2)6/h3,5H,1-2H3
Formula Weight 88.0524
Chemical Name or Material Acetoin
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.