Learn More
Acetohydrazide-D3, TRC
Specifications
Specifications
| Chemical Name or Material | Acetohydrazide-D3 |
| CAS | 1028333-41-6 |
| Molecular Formula | C2H3D3N2O |
| Quantity | 100 mg |
| Synonym | Acetic-2,2,2-D3 Acid Hydrazide |
| InChI Formula | InChI=1S/C2H6N2O/c1-2(5)4-3/h3H2,1H3,(H,4,5)/i1D3 |
| SMILES | O=C(C([2H])([2H])[2H])NN |
| IUPAC Name | 2,2,2-trideuterioacetohydrazide |
| Molecular Weight (g/mol) | 77.1 |