Learn More
Acetohexamide, TRC
Product Code. 30180278
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180278 1 g 1g
30180002 100 mg 100mg
30180419 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30180278 Supplier Toronto Research Chemicals Supplier No. TRCA1626501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 968-81-0
Molecular Formula C15 H20 N2 O4 S
Molecular Weight (g/mol) 324.4
Synonym Benzenesulfonamide, 4-acetyl-N-[(cyclohexylamino)carbonyl]-Urea, 1-[(p-acetylphenyl)sulfonyl]-3-cyclohexyl- (6CI,8CI), 1-(p-Acetylbenzenesulfonyl)-3-cyclohexylurea, 1-[(p-Acetylphenyl)sulfonyl]-3-cyclohexylurea, Acetohexamid, Acetohexamide, Dimelin, Dimelin (antidiabetic), Dimelor, Dymelor, Gamadiabet, Hypoglicil, Metaglucina, Minoral, N-(p-Acetylphenylsulfonyl)-N'-cyclohexylurea, Ordimel, Tsiklamid, U 14812
IUPAC Name 1-(4-acetylphenyl)sulfonyl-3-cyclohexylurea
SMILES CC(=O)c1ccc(cc1)S(=O)(=O)NC(=O)NC2CCCCC2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C15H20N2O4S/c1-11(18)12-7-9-14(10-8-12)22(20,21)17-15(19)16-13-5-3-2-4-6-13/h7-10,13H,2-6H2,1H3,(H2,16,17,19)
Formula Weight 324.1144
Percent Purity >95%
Chemical Name or Material Acetohexamide
Grade HPLC