Learn More
Acetohexamide-d11, TRC
Specifications
Specifications
| Chemical Name or Material | Acetohexamide-D11 (cyclohexyl-D11) |
| CAS | 2468796-77-0 |
| Molecular Formula | C15 H9 D11 N2 O4 S |
| Quantity | 1 mg |
| Synonym | Acetohexamide-D11, Acetohexamide D11 |
| InChI Formula | InChI=1S/C15H20N2O4S/c1-11(18)12-7-9-14(10-8-12)22(20,21)17-15(19)16-13-5-3-2-4-6-13/h7-10,13H,2-6H2,1H3,(H2,16,17,19)/i2D2,3D2,4D2,5D2,6D2,13D |
| SMILES | [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(NC(=O)NS(=O)(=O)c2ccc(cc2)C(=O)C)C([2H])([2H])C1([2H])[2H] |
| IUPAC Name | 1-(4-acetylphenyl)sulfonyl-3-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)urea |
| Molecular Weight (g/mol) | 335.46308 |
| Formula Weight | 335.1834 |