Learn More
Acetochlor-ethane sulfonic acid (ESA) sodium 100 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30327395
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327395 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327395 Supplier Dr. Ehrenstorfer Supplier No. DREA10018210ME100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acetochlor Ethane Sulfonic Acid Sodium Salt
CAS 947601-84-5
Analyte or Component Names Acetochlor Ethane Sulfonic Acid Sodium Salt
Shipping Condition +20°C
Molecular Formula C14 H20 N O5 S . Na
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C14H21NO5S.Na/c1-4-12-8-6-7-11(3)14(12)15(10-20-5-2)13(16)9-21(17,18)19;/h6-8H,4-5,9-10H2,1-3H3,(H,17,18,19);/q;+1/p-1
SMILES [Na+].CCOCN(C(=O)CS(=O)(=O)[O-])c1c(C)cccc1CC
IUPAC Name sodium;2-[N-(ethoxymethyl)-2-ethyl-6-methylanilino]-2-oxoethanesulfonate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 337.37
Quantity 1 mL
Formula Weight 337.096 g/mol