missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, TRC

Product Code. 30008552
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Unit Size:
1 L
2.5 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008552 2.5 L 2.5 L
30003600 5 L 5 L
17943705 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008552 Supplier Toronto Research Chemicals Supplier No. TRCA1676402.5L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2 H4 O2
Molecular Weight (g/mol) 60.05
Synonym PN: WO2017048620 SEQID: 12 claimed sequence, Aci-Jel, E 260, Engage Agro, Ethanoic acid, Ethanoic acid monomer, Ethylic acid, Glacial acetic acid, HACP 85, Methanecarboxylic acid, Munger Horticultural Vinegar Plus, NSC 111201, NSC 112209, NSC 115870, NSC 127175, NSC 132953, NSC 406306, Vinegar acid, Weed Pharm, acetic acid
IUPAC Name acetic acid
SMILES CC(=O)O

Specifications

InChI Formula InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
Formula Weight 60.0211
Quantity 2.5 L
Chemical Name or Material Acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.