Learn More
Acetanilide, Sigma-Aldrich™
Product Code. 15834681
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15834681 500 g 500g
15824681 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15834681 Supplier Sigma Aldrich Supplier No. 8223440500
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103-84-4
Grade Research
Molecular Weight (g/mol) 135.16
Specifications
Melting Point 115°C
Density 1.22 g/cm3 at 20°C
Boiling Point 304°C/1013 hPa
MDL Number MFCD00008674
Quantity 500 g
Synonym Acetanilide, N-Phenylacetamide, Acetanilide