Learn More
Acetamiprid, TRC
Product Code. 18053710
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18053710 500 mg 500mg
18020820 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18053710 Supplier Toronto Research Chemicals Supplier No. TRCA150800500MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 135410-20-7
Molecular Formula C10 H11 Cl N4
Molecular Weight (g/mol) 222.67
Synonym (1E)-N-[(6-Chloro-3-pyridinyl)methyl]-N'-cyano-N-methylethanimidamide, ADA 06200, Acetamiprid, Assail, Assail 30SG, BY 102960, Epik 20 SG, Gazelle SG, Intruder, Mospilan, Mospilan 20SG, Mospilan 20SP, Mospilan SG, Mukhnet A, NFK 17, NI 25, NI 25 (pesticide), Piorun, Pristine, Pristine (insecticide), Prize, Prize (insecticide), Quasar 8.5SL, Stonkat, Supreme, TD 2472, TD 2472-01, TD 2480
IUPAC Name N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
SMILES CN(Cc1ccc(Cl)nc1)\C(=N\C#N)\C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C10H11ClN4/c1-8(14-7-12)15(2)6-9-3-4-10(11)13-5-9/h3-5H,6H2,1-2H3/b14-8+
Formula Weight 222.0672
Percent Purity >95%
Chemical Name or Material Acetamiprid
Grade HPLC