missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen Dimer, TRC

Product Code. 17980815
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980815 25 mg 25mg
17903545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17980815 Supplier Toronto Research Chemicals Supplier No. TRCA16123525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98966-14-4
Molecular Formula C16H16N2O4
Molecular Weight (g/mol) 300.31
Synonym Acetamide, N,N'-(6,6'-dihydroxy[1,1'-biphenyl]-3,3'-diyl)bis-, N,N'-(6,6'-Dihydroxy[1,1'-biphenyl]-3,3'-diyl)bis[acetamide], N-[3-(5-Acetamido-2-hydroxyphenyl)-4-hydroxyphenyl]acetamide, Acetaminophen Dimer, Diparacetamol, N,N'-(6,6'-Dihydroxy[1,1'-biphenyl]-3,3'-diyl)bis-acetamide, 2,2'-Dihydroxy-5,5'-diacetyldiaminebiphenyl
IUPAC Name N-[3-(5-acetamido-2-hydroxyphenyl)-4-hydroxyphenyl]acetamide
SMILES CC(=O)Nc1ccc(O)c(c1)c2cc(NC(=O)C)ccc2O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16N2O4/c1-9(19)17-11-3-5-15(21)13(7-11)14-8-12(18-10(2)20)4-6-16(14)22/h3-8,21-22H,1-2H3,(H,17,19)(H,18,20)
Formula Weight 300.111
Chemical Name or Material Acetaminophen Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.