Learn More
Acetamidine hydrochloride, 97%
Product Code. 11473107
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11473107 500 g 500g
11384619 25 g 25g
11463107 100 g 100g
3 options
Product Code. 11473107
Brand: Thermo Scientific Alfa Aesar A16091.36
Request Bulk or Custom Format
CAS: 124-42-5 | C2H7ClN2 | 94.542 g/mol
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.
Solubility
Soluble in water.
Notes
It is hygroscopic. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 124-42-5
Molecular Formula C2H7ClN2
Molecular Weight (g/mol) 94.542
MDL Number MFCD00013016
InChI Key WCQOBLXWLRDEQA-UHFFFAOYSA-N
Synonym acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
PubChem CID 67170
IUPAC Name ethanimidamide;hydrochloride
SMILES CC(=N)N.Cl
Specifications
Melting Point 168°C to 175°C
Quantity 500 g
Beilstein 3591762
Sensitivity Hygroscopic
Merck Index 14,44
Solubility Information Soluble in water.
Formula Weight 94.54
Percent Purity 97%
Chemical Name or Material Acetamidine hydrochloride