Learn More
Acetaldoxime, Sigma-Aldrich™
Specifications
Specifications
| CAS | 107-29-9 |
| Density | 0.969 g/cm3 at 20°C |
| MDL Number | MFCD00002124 |
| Quantity | 50 mL |
| Synonym | Acetaldoxime, Acetaldehyde oxim, Acetaldoxime |
| Molecular Weight (g/mol) | 59.07 |
| Grade | Research |