missing translation for 'onlineSavingsMsg'
Learn More

Acetaldoxime, Sigma-Aldrich™

Product Code. 32238242
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238242 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238242 Supplier Sigma Aldrich Supplier No. 8410850250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

(Mixture of cis and trans isomers) for synthesis

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetaldoxime
CAS 107-29-9
Density 0.969 g/cm³ at 20°C
Flash Point 40°C - open cup)
Quantity 250 mL
Synonym Acetaldoxime, Acetaldehyde oxim, Acetaldoxime
Molecular Weight (g/mol) 59.07
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.