Learn More
Acetaldehyde, Sigma-Aldrich™
Description
Acetaldehyde is an organic compound and one of the significant aldehydes. Acetaldehyde is employed to produce acetic anhydride, acetic acid, cellulose acetate, pyridine derivatives, pentaerythritol, peracetic acid, and terephthalic acid. Furthermore, it is used as a solvent.
Specifications
Specifications
| Chemical Name or Material | Acetaldehyde |
| CAS | 75-07-0 |
| Melting Point | -123°C |
| Density | 0.78 g/cm³ at 20°C |
| Boiling Point | 20.4°C (1013 hPa) |
| Flash Point | -38.89°C (closed cup) |
| Quantity | 500 mL |
| Synonym | Acetaldehyde, Ethanal, Acetic Aldehyde, Ethyl Aldehyde, Acetaldehyde |
| Molecular Weight (g/mol) | 44.05 |
| Grade | Not Available |