Learn More
Fisherbrand™ accumet™ AB315 and AB330 Meter Replacement Universal Power Adapter
Artikelnummer. 17358949 Alle ansehen Fisherbrand Traceable Produkte
Ansicht ändern
Click to view available options
For Use With (Equipment):
accumet AB315 and AB330 Meters
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. For Use With (Equipment) Packungsgröße
17358949 accumet AB315 and AB330 Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17358949
  • ONLINE EXCLUSIVE PRICE
    52.35 EUR
    38.09 EUR / Pack of 1
    Save 14.26 EUR
    27% Off
Estimated Shipment: 15-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Select accessories for your Fisherbrand™ accumet™ AB315 pH/mV meter and Fisherbrand™ accumet™ AB330 conductivity meter.
  • Universal 100/240 V 50/60 Hz power adapter
  • Includes wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China
TRUSTED_SUSTAINABILITY
Spezifikation
Type Replacement universal power adapter
Frequency 50/60 Hz
Includes Wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China
Voltage 100/240 V
Compatibility accumet™ AB300 series meters
For Use With (Equipment) accumet AB315 and AB330 Meters