Learn More
Fisherbrand™ AccuDuo Cartridge
Product Code. 15459719 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
H Pack
P Pack
U Pack
U Pack (low TOC)
U Pack w/ 0.2μm PES Capsule Filter
For Use With (Equipment):
Accu20
Accu20, Accu500
Accu500
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) unitSize
15459719 H Pack Accu500 Each
15449719 P Pack Accu20 Each
15489729 U Pack Accu20, Accu500 Each
15499729 U Pack w/ 0.2μm PES Capsule Filter Accu20, Accu500 Each
15409739 U Pack (low TOC) Accu20, Accu500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15459719
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
  • Includes two sets of dual-column purification cartridges
  • Cartridges work sequentially to ensure thorough removal of ions and organics in water
Specifications
For Use With (Equipment) Accu500
Type H Pack
For Use With (Application) Ultrapure water for critical laboratory applications