Learn More
BRAND™ Accu-jet™ Pro Snap Tapes
Product Code. 10563872 Shop All Brand Products
Change view
Click to view available options
Dimensions (L x W):
12 x 65 mm
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10563872 12 x 65 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10563872 Supplier BRAND™ Supplier No. 26542
  • ONLINE EXCLUSIVE PRICE
    8.75 EUR
    8.21 EUR / Pack of 2
    Save 0.54 EUR
    6% Off
View all promotions
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Bulk Quote Request
2 sets for accu-jet™ pro pipette controller
TRUSTED_SUSTAINABILITY
Specifications
Description Snap Tapes
Dimensions (L x W) 12 x 65 mm
Includes 2 sets
For Use With (Equipment) Accu-jet Pro Pipet Controller