Learn More
Thermo Scientific™ Replacement HEPA Filter
Product Code. 15363022 Shop All Thermo Scientific Products
Change view
Click to view available options
Features:
99.95% Efficiency at MPPS
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Features Unit Size
15363022 99.95% Efficiency at MPPS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15363022 Supplier Thermo Scientific™ Supplier No. 50141920
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
For Use With: Heracell VIOS 160i/250i and Forma Steri-cycle i160/i250
Help ensure measurable Class 10 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.95% separation efficiency @MPPS
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement in chamber HEPA filter, rectangular
For Use With (Equipment) Heracell VIOS 160i/250i and Forma Steri-cycle i160/i250
Lighting LED
Type HEPA Filter
Features 99.95% Efficiency at MPPS