missing translation for 'onlineSavingsMsg'
Learn More

Acalabrutinib, MedChemExpress

Product Code. 16497408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497408 10 mg 10mg
16489887 5 mg 5mg
16487386 100 mg 100mg
16463437 50 mg 50mg
16441276 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497408 Supplier Medchem Express Supplier No. HY17600
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Acalabrutinib (ACP-196) is an orally active, irreversible, and highly selective second-generation BTK inhibitor. Acalabrutinib binds covalently to Cys481 in the ATP-binding pocket of BTK. Acalabrutinib demonstrates potent on-target effects and efficacy in mouse models of chronic lymphocytic leukemia (CLL).

Acalabrutinib (ACP-196) is an orally active, irreversible, and highly selective second-generation BTK inhibitor. Acalabrutinib binds covalently to Cys481 in the ATP-binding pocket of BTK. Acalabrutinib demonstrates potent on-target effects and efficacy in mouse models of chronic lymphocytic leukemia (CLL).

Specifications

Chemical Name or Material Acalabrutinib
CAS 1420477-60-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H23N7O2
Quantity 10 mg
Synonym ACP-196
Solubility Information DMSO : 250 mg/mL (537.05 mM; Need ultrasonic)
SMILES O=C(NC1=NC=CC=C1)C2=CC=C(C3=C4C(N)=NC=CN4C([C@H]5N(C(C#CC)=O)CCC5)=N3)C=C2
Molecular Weight (g/mol) 465.51
Formula Weight 465.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.