missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AC 265347

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AC 265347 is a CaSR biased allosteric modulator (pEC50 = 7.8-8.1). Calcimimetic. Displays ability to bias signalling towards the accumulation of pERK1/2 and IP1. Reduces parathyroid hormone (PTH) levels in rat serum without inducing the release of calcitonin. Exhibits no significant activation of human GABAB or type I PTH receptors. Orally active.
Specifications
Specifications
| Chemical Name or Material | α-(2,4-Dimethylphenyl)-α-methyl-2-benzothiazolemethanol |
| CAS | 1253901-26-6 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Molecular Formula | C17H17NOS |
| Formula Weight | Observed MW: 283.39 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction