missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AC 264613
GREENER_CHOICE

Product Code. p-4783707
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025282

Brand: Tocris Bioscience™ 3370/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PAR2 receptor agonist

Potent and selective protease-activated receptor 2 (PAR2) agonist (pEC50 = 7.5). Displays no activity at other PAR subtypes and exhibits no significant activity at over 30 other receptors implicated in nociception and inflammation. Stimulates PI hydrolysis, Ca2+ mobilization and cellular proliferation in vitro (pEC50 values are 6.9, 7.0 and 7.5 respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1051487-82-1
Molecular Formula C19H18BrN3O2

Specifications

Quantity 10 mg
Formula Weight 400.27
Purity >98%
Molecular Weight (g/mol) 400.276
Chemical Name or Material AC 264613
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.