Learn More
Testo™ ABS Sound Level Measuring Instrument
Product Code. 10358303
Change view
Click to view available options
Length (Metric):
255 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10358303 255 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10358303 Supplier Testo™ Supplier No. 0563 8155
  • ONLINE EXCLUSIVE PRICE
    834.75 EUR
    300.00 EUR / Pack of 1
    Save 534.75 EUR
    64% Off
Estimated Shipment: 29-12-2026
Specifications
Battery Type 9 V Block Battery, 6F22
Dimensions 255 x 55 x 43 mm
For Use With (Application) Determine the noise level quickly, easily, and reliably in day-to-day work
Height (English) 2 in.
Height (Metric) 43 mm
Includes Adjustment screwdriver, Integrated tripod knuckle screw (0.25 in.)
Length (English) 10 in.
Length (Metric) 255 mm
Measurement Range 32 to 130 dB
Memory Min./max. memory
Resolution 0.1 dB
Response Time 5 sec.
Weight (English) 7 oz.
Weight (Metric) 195 g
Width (English) 2 in.
Width (Metric) 55 mm