Learn More
Abcam Rat monoclonal [SB77e] Anti-Mouse IgG1 H&L (Alkaline Phosphatase) preadsorbed, 500 uL
Product Code. 32109168 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109168 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32109168 Supplier Abcam Supplier No. ab99602500uL
 Restricted Item
Estimated Shipment: 25-11-2026
Rat Monoclonal antibody, suitable for Flow Cyt, ELISA, with 2 citations.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Applications Flow Cytometry
Classification Monoclonal
Clone SB77e
Conjugate Unconjugated
Formulation pH: 8, Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 0.605% Tris, 0.0095% Magnesium chloride
Gene Ighg1
Gene Symbols Ighg1
Host Species Rat
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Ighg1, Mouse
Validated Application abrv Flow Cyt, ELISA