Learn More
Abcam Human/Monkey TNF alpha Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32115305 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115305 10 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32115305 Supplier Abcam Supplier No. ab24179110x96Tests
 Restricted Item
Estimated Shipment: 12-11-2026
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Human, Rhesus monkey and Cynomolgus monkey
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols TNF
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human, Rhesus monkey and Cynomolgus monkey.
Target Species Human, Rhesus Monkey, Cynomolgus Monkey
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu, RhMk
Validated Application abrv sELISA