Learn More
Atago™ Abbe DR-A1 Refractometer
Product Code. 10041132
Change view
Click to view available options
Dimensions (L x W x H):
130 x 290 x 310 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10041132 130 x 290 x 310 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10041132 Supplier Atago™ Supplier No. 1310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Refractometer
Refractive Index 1.3 to 1.7
Accuracy ±0.0002nD
Display Digital
Includes With AC adapter
Quantity 1 Each
Dimensions (L x W x H) 130 x 290 x 310 mm
Height (Metric) 310 mm
Length (Metric) 130 mm
Width (Metric) 290 mm
Temperature Range (Metric) 5°C to 50°C
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz