Learn More
3M™ Multi-Format Chemical Sorbent
Product Code. 12357979
Change view
Click to view available options
Capacity (Metric):
119 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12357979 119 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12357979 Supplier 3M™ Supplier No. 7000002024
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Effectively controls aggressive liquid spills including acids and alkalis. 3M™ Multi-format Chemical Sorbent combines four formats in one product; mini-boom, pillow, sheet and roll.
Effective clean up and control of aggressive liquid spills including acids and alkalis. The dispenser combines four formats in one product; mini-boom, pillow, sheet and roll.
  • Sheets are perforated at 40cm intervals
  • Can be used with many aggressive chemicals
  • Immediate response to chemical spillages for fast and effective containment and clean-up
  • Sorbent materials can be disposed of easily.
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Capacity (Metric) 119 L
Type Multi-format Chemical Sorbents
For Use With (Application) Cleaning and containing chemical-based spills such as acids/alkalis/alcohols, chemicals, flammable liquids, and other solvents.
Packaging Quantity 1 Ea.
Length (Metric) 15.2 m
Width (Metric) 120 mm
Weight (Metric) 8.2 kg