Learn More
AASS Antibody - Azide and BSA Free, Novus Biologicals™
Product Code. 18664271 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18664271 0.02 mL 0.02mL
18619930 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18664271 Supplier Novus Biologicals Supplier No. NBP2920930.02ml
Rabbit Polyclonal Antibody
AASS Polyclonal antibody specifically detects AASS in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen AASS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias aminoadipate-semialdehyde synthase, aminoadipic semialdehyde synthase, LKR/SDH, LKRSDH, LORSDH, lysine-2-oxoglutarate reductase, mitochondrial
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-400 of human AASS (NP_005754.2). MHIGMAHNYRNSSQAVQAVRDAGYEISLGLMPKSIGPLTFVFTGTGNVSKGAQAIFNELPCEYVEPHELKEVSQTGDLRKVYGTVLSRHHHLVRKTDAVYDPAEYDKHPERYISRFNTDIAPYTTCLINGIYWEQNTPRLLTRQDAQSLLAPGKFSPAGVEGCPALPHKLVAICDISADTGGSIEFMTECTTIEHPFCMYDADQHIIHDSVEGSGILMCSI
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10157
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG