Learn More
a-Zingiberene, TRC
Product Code. 30455589
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455589 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455589 Supplier Toronto Research Chemicals Supplier No. TRCZ4399800.5MG
 Restricted Item
Estimated Shipment: 10-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material alpha-Zingiberene
CAS 495-60-3
Molecular Formula C15 H24
Quantity 0.5 mg
Synonym 1,3-Cyclohexadiene','5-[(1S)-1,5-dimethyl-4-hexen-1-yl]-2-methyl-','(5R)-, 1,3-Cyclohexadiene','5-(1,5-dimethyl-4-hexenyl)-2-methyl-','[S-(R*,S*)]-, 1,3-Cyclohexadiene','5-[(1S)-1,5-dimethyl-4-hexenyl]-2-methyl-','(5R)- (9CI), Zingiberene (7CI), (5R)-5-[(1S)-1,5-Dimethyl-4-hexen-1-yl]-2-methyl-1,3-cyclohexadiene, (-)-Zingiberene, l-Zingiberene, α-Sesquiphellandrene, α-Zingiberene
InChI Formula InChI=1S/C15H24/c1-12(2)6-5-7-14(4)15-10-8-13(3)9-11-15/h6,8-10,14-15H,5,7,11H2,1-4H3/t14-,15+/m0/s1
SMILES C[C@@H](CCC=C(C)C)[C@H]1CC=C(C)C=C1
IUPAC Name (5R)-2-methyl-5-[(2S)-6-methylhept-5-en-2-yl]cyclohexa-1,3-diene
Molecular Weight (g/mol) 204.35
Formula Weight 204.1878