Learn More
A-966492, MedChemExpress
Product Code. 16486767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486767 5 mg 5mg
16464068 10 mg 10mg
16420427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16486767 Supplier Medchem Express Supplier No. HY10614
MedChemExpress A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1.5 nM, respectively.
A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1.5 nM, respectively.
Specifications
Chemical Name or Material A-966492
CAS 934162-61-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H17FN4O
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (308.31 mM)
SMILES FC1=CC([C@H]2NCCC2)=CC=C1C3=NC4=C(C(N)=O)C=CC=C4N3
Molecular Weight (g/mol) 324.35
Formula Weight 324.35
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death