missing translation for 'onlineSavingsMsg'
Learn More

A 922500, MedChemExpress

Product Code. 16465717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465717 5 mg 5mg
16449546 50 mg 50mg
16442978 10 mg 10mg
16424296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465717 Supplier Medchem Express Supplier No. HY10038

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A 922500 (DGAT-1 Inhibitor 4a) is a potent, selective, and orally bioavailable diacylglycerol acyltransferase 1 (DGAT-1) inhibitor with IC50s of 9 and 22 nM against human and mouse DGAT-1, respectively.

A 922500 (DGAT-1 Inhibitor 4a) is a potent, selective, and orally bioavailable diacylglycerol acyltransferase 1 (DGAT-1) inhibitor with IC50s of 9 and 22 nM against human and mouse DGAT-1, respectively.

Specifications

Chemical Name or Material A 922500
Physical Form Solid
CAS 959122-11-3
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C26H24N2O4
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym DGAT-1 Inhibitor 4a
Solubility Information DMSO : ≥ 50 mg/mL (116.69 mM)
SMILES O=C(NC1=CC=C(C2=CC=C(C([C@@H]3CCC[C@H]3C(O)=O)=O)C=C2)C=C1)NC4=CC=CC=C4
Molecular Weight (g/mol) 428.48
Formula Weight 428.48
Percent Purity 95.43%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.