Description
- ANSI-compliant footprint for compatibility with automated systems
- Free of detectable DNase, RNase, and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | DIN ISO 9001 |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Show More |