Learn More
Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural
Product Code. 12154112 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12154112 Natural Pack of 25
12194112 Yellow Pack of 25
12184112 Green Pack of 25
12164112 Red Pack of 25
12174112 Blue Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12154112
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
Bulk Quote Request
Fit most thermal cyclers
  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Stackable
  • Raised-rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 200μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • SBS (Society for Biomolecular Screening) footprint
  • Automation friendly
  • Eight holes in the skirt aid plate positioning and removal from thermal cycler block
  • Cleanroom produced
Specifications
Format Standard
Array 8 x 12
Color Natural
Product Type PCR Microplate
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes