Learn More
9-Phenylacridine, Thermo Scientific™
Product Code. 17770709 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17770709 1 g 1g
17780709 5 g 5g
17790709 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17770709 Supplier Thermo Scientific Acros Supplier No. 469430010
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Specifications
Alpha Vector PHENYLACRIDINE
Chemical Name or Material 9-Phenylacridine
CAS 602-56-2
Melting Point 183°C to 187°C
Color Yellow
Boiling Point 411°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C19 H13 N
MDL Number 00092562
Quantity 1 g
Molecular Weight (g/mol) 255.32
Physical Form Crystalline Powder