missing translation for 'onlineSavingsMsg'
Learn More

9-Phenanthrol, TRC

Product Code. 30007317
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007317 100 mg 100mg
30010708 1 g 1g
30004349 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007317 Supplier Toronto Research Chemicals Supplier No. TRCP295020100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 484-17-3
Molecular Formula C14 H10 O
Molecular Weight (g/mol) 194.23
Synonym 9-Phenanthrenol (9CI, ACI), 9-Phenanthrol (6CI, 7CI, 8CI), 9-Hydroxyphenanthrene, NSC 50554
IUPAC Name phenanthren-9-ol
SMILES Oc1cc2ccccc2c3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H10O/c15-14-9-10-5-1-2-6-11(10)12-7-3-4-8-13(12)14/h1-9,15H
Formula Weight 194.0732
Percent Purity >95%
Chemical Name or Material 9-Hydroxy-phenanthrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.