Learn More
9-cis-Retinoic Acid, >98%, MP Biomedicals™
Product Code. 11419931 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419931 5 mg 5mg
11409931 1 mg 1mg
11429931 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11419931 Supplier MP Biomedicals Supplier No. 0215872505
  • ONLINE EXCLUSIVE PRICE
    542.00 EUR
    515.55 EUR / 5mg
    Save 26.45 EUR
    4% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Chemical Identifiers
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
ChEBI CHEBI:50648
Specifications
CAS 5300-03-8
Assay Percent Range ≥98%
Quantity 5 mg
Synonym alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
IUPAC Name (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
PubChem CID 449171
Formula Weight 300.44
Physical Form Powder
Chemical Name or Material 9-cis-Retinoic Acid, Cell Biology and Analysis