Learn More
9,9-Dimethyl-9,10-dihydroacridine, TRC
Specifications
Specifications
| CAS | 6267-02-3 |
| Molecular Formula | C15H15N |
| Quantity | 250 mg |
| Synonym | 9,9-Dimethylacridan, 9,10-Dihydro-9,9-dimethylacridine, 9,9-Dimethyl-10H-acridine, NSC 36671 |
| SMILES | CC1(C)c2ccccc2Nc3ccccc13 |
| Molecular Weight (g/mol) | 209.29 |
| Formula Weight | 209.1204 |
| Percent Purity | >95% |
| Chemical Name or Material | 9,9-Dimethyl-9,10-dihydroacridine |
| Grade | HPLC |