Learn More
9-(4-Acetoxy-3-acetoxymethylbutyl)-2-amino-6-chloropurine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 9-(4-Acetoxy-3-acetoxymethylbutyl)-2-amino-6-chloropurine-d4 |
| CAS | 1020718-81-3 |
| Molecular Formula | C14 2H4 H14 Cl N5 O4 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C14H18ClN5O4/c1-8(21)23-5-10(6-24-9(2)22)3-4-20-7-17-11-12(15)18-14(16)19-13(11)20/h7,10H,3-6H2,1-2H3,(H2,16,18,19)/i3D,4D,5D,6D |
| SMILES | [2H]C(OC(=O)C)C(C([2H])OC(=O)C)C([2H])C([2H])n1cnc2c(Cl)nc(N)nc12 |
| IUPAC Name | [2-[acetyloxy(deuterio)methyl]-4-(2-amino-6-chloropurin-9-yl)-1,3,4-trideuteriobutyl] acetate |
| Molecular Weight (g/mol) | 359.8 |
| Formula Weight | 359.1298 |