Learn More
9,11-Didehydrooestriol, TRC
Specifications
Specifications
| Chemical Name or Material | 9,11-Didehydroestriol |
| CAS | 246021-20-5 |
| Molecular Formula | C18 H22 O3 |
| Quantity | 25 mg |
| Synonym | Estra-1,3,5(10),9(11)-tetraene-3,16alpha,17beta-triol, Estriol Imp. A (EP), 9,11-Didehydroestriol |
| InChI Formula | InChI=1S/C18H22O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5-6,8,14-17,19-21H,2,4,7,9H2,1H3/t14-,15+,16-,17+,18+/m1/s1 |
| SMILES | C[C@]12CC=C3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O |
| IUPAC Name | (8S,13S,14S,16R,17R)-13-methyl-6,7,8,12,14,15,16,17-octahydrocyclopenta[a]phenanthrene-3,16,17-triol |
| Molecular Weight (g/mol) | 286.37 |
| Formula Weight | 286.1569 |