Learn More
Axygen™ 80-Well Microcentrifuge Tube Rack
Product Code. 12587847 Shop All Axygen Scientific Products
Change view
Click to view available options
Array:
5 x 16
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Array unitSize
12587847 5 x 16 Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12587847 Supplier Axygen™ Supplier No. R80R
  • ONLINE EXCLUSIVE PRICE
    158.00 EUR
    156.01 EUR / Pack of 25
    Save 1.99 EUR
    1% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
This 80-well polypropylene microtube rack holds 1.5/2.0mL screw cap microtubes, making it an excellent option for cryogenic storage.
  • Red in color
  • Conical bottom wells offer superior stability for conical tubes
  • Each well is numbered for ease of sample identification
  • Includes a frosted side writing panel
  • Resistant to most lab chemicals and solvents
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Holds Screw Cap Microtubes (1.5 to 2 mL)
Material Polypropylene
Positions 80
For Use With (Application) Storage of cryogenic tubes
Array 5 x 16